Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171653.1 | NP_001165124.1 | 7 | 2102 | ||
NM_014795.3 | NP_055610.1 | 8 | 2174 | |||
XM_006712881.3 | XP_006712944.1 | 8 | 1768 | |||
XM_006712882.3 | XP_006712945.1 | 8 | 1817 | |||
XM_011512232.2 | XP_011510534.1 | 7 | 1811 | |||
XM_017005414.1 | XP_016860903.1 | 9 | 1888 | |||
XM_017005415.1 | XP_016860904.1 | 7 | 2791 | |||
GenBank mRNA | AB011141.1 | - | 8 | 2096 | ||
AB056507.1 | - | 8 | 2131 | |||
AB193095.1 | - | 2032 | ||||
AB193096.1 | - | 7 | 1657 | |||
AK294928.1 | - | 7 | 1830 | |||
AK308445.1 | - | 8 | 1781 | |||
BC127101.1 | - | 8 | 1786 | 40124119 | ||
BC127102.1 | - | 8 | 1785 | 40124124 |