Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018078.2 | NP_001018088.1 | 9, 10 | 907 | ||
NM_001288803.1 | NP_001275732.1 | 8, 9 | 810 | |||
NM_004957.5 | NP_004948.4 | 9, 10 | 888 | |||
NR_110170.1 | 9, 10 | 869 | ||||
XM_005251864.3 | XP_005251921.1 | 9, 10 | 847 | |||
XM_011518439.2 | XP_011516741.1 | 6, 7 | 521 | |||
XM_017014565.1 | XP_016870054.1 | 9, 10 | 907 | |||
XM_017014566.1 | XP_016870055.1 | 9, 10 | 845 | |||
GenBank mRNA | AK056920.1 | - | 9, 10 | 817 | ||
AK295309.1 | - | 8, 9 | 786 | |||
BC007121.1 | - | 9, 10 | 819 | |||
BC064393.1 | - | 9, 10 | 850 | 5750645 | ||
BG328181.1 | - | 46 | ||||
M98045.1 | - | 9, 10 | 765 |