Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145128.2 | NP_001138600.2 | 26 | 3167 | ||
XM_006715376.3 | XP_006715439.2 | 27 | 4032 | |||
XM_011535550.2 | XP_011533852.1 | 27 | 4032 | |||
XM_011535552.2 | XP_011533854.1 | 26 | 3208 | |||
XM_011535553.2 | XP_011533855.1 | 26 | 3815 | |||
XM_011535554.2 | XP_011533856.1 | 25 | 3711 | |||
XM_011535555.2 | XP_011533857.1 | 27 | 4032 | |||
XM_011535556.2 | XP_011533858.1 | 27 | 4032 | |||
XM_011535558.1 | XP_011533860.1 | 8 | 1053 | |||
XM_017010384.1 | XP_016865873.1 | 27 | 4032 | |||
XM_017010385.1 | XP_016865874.1 | 26 | 3200 | |||
XM_017010386.1 | XP_016865875.1 | 27 | 4032 | |||
XM_017010387.1 | XP_016865876.1 | 21 | 3663 | |||
XM_017010388.1 | XP_016865877.1 | 27 | 4032 | |||
XM_017010389.1 | XP_016865878.1 | 12 | 1915 | |||
GenBank mRNA | AK131244.1 | - | 2 | 575 |