Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212558.2 | NP_997723.2 | 2 | 981 | ||
GenBank mRNA | AK125320.1 | - | 2 | 981 | ||
BX647550.1 | - | 2 | 940 | |||
CR749589.1 | - | 1 | 802 |