Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR14I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004734.1 | NP_001004734.1 | 1 | 209 | ||
XM_005273132.3 | XP_005273189.1 | 2 | 344 | |||
GenBank mRNA | BC137188.1 | - | 1 | 233 | 9021185 | |
BC137189.1 | - | 1 | 233 | 9021186 |