Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212559.2 | NP_997724.2 | 3 | 1851 | ||
XM_005262130.2 | XP_005262187.1 | 2 | 2170 | |||
XM_011530955.1 | XP_011529257.1 | 4 | 1219 | |||
XM_017029517.1 | XP_016885006.1 | 2 | 945 | |||
GenBank mRNA | AK296711.1 | - | 2 | 2106 | ||
AY534239.1 | - | 3 | 1423 | |||
AY589511.1 | - | 3 | 1851 | |||
BC137010.1 | - | 3 | 1493 | 9021003 | ||
BC137011.1 | - | 3 | 1493 | 9021004 |