Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099270.1 | NP_001092740.1 | 2 | 1210 | ||
XM_005251989.4 | XP_005252046.1 | 3 | 1323 | |||
XM_011518699.2 | XP_011517001.1 | 2 | 1226 | |||
GenBank mRNA | AB082524.1 | - | 2 | 1210 | ||
DQ227306.1 | - | 1 | 1125 |