Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207581.3 | NP_997464.2 | 1, 2 | 437 | ||
XM_017022180.1 | XP_016877669.1 | 1, 2 | 441 | |||
GenBank mRNA | AK303955.1 | - | 1, 2 | 436 | ||
BC137465.1 | - | 1, 2 | 358 | 9021465 | ||
DC393748.1 | - | 1, 2 | 437 | |||
DQ489734.1 | - | 1, 2 | 288 |