Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002438.3 | NP_002429.1 | 5 | 964 | ||
GenBank mRNA | AK308132.1 | - | 5 | 965 | ||
AK308167.1 | - | 5 | 960 | |||
BC142642.1 | - | 5 | 945 | |||
BC146838.1 | - | 5 | 887 | 9056765 | ||
DQ663787.1 | - | 5 | 841 | |||
J05550.1 | - | 5 | 944 | |||
X55635.1 | - | 5 | 917 |