Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001852.3 | NP_001001852.2 | 6 | 1665 | ||
GenBank mRNA | AB114795.1 | - | 6 | 1647 | ||
AK292005.1 | - | 6 | 1313 | |||
BC017083.2 | - | 1 | 422 | |||
BC052239.1 | - | 6 | 1246 | |||
BC064477.1 | - | 2 | 638 | |||
BC141855.1 | - | 6 | 1278 | 8860375 | ||
CT000425.1 | - | 2 | 465 |