Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM41B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165030.2 | NP_001158502.1 | 2 | 467 | ||
NM_015012.3 | NP_055827.1 | 2 | 467 | |||
NR_028491.2 | 2 | 467 | ||||
GenBank mRNA | AK303339.1 | - | 2 | 158 | ||
BC035034.1 | - | 2 | 338 | 5259179 | ||
BC044597.1 | - | 2 | 329 | 4811565 | ||
BC064366.1 | - | 2 | 224 | |||
BC091524.1 | - | 2 | 266 | 30512350 | ||
BI596931.1 | - | 2 | 471 | |||
BI829492.1 | - | 2 | 447 | |||
D26067.1 | - | 2 | 273 | |||
DA149513.1 | - | 2 | 333 | |||
DR157551.1 | - | 2 | 320 |