Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 16 | 2301 | ||
NR_027410.1 | 24 | 4549 | ||||
XM_011521596.1 | XP_011519898.1 | 23 | 3146 | |||
XM_011521597.1 | XP_011519899.1 | 22 | 3097 | |||
XM_017022210.1 | XP_016877699.1 | 24 | 3450 | |||
XM_017022211.1 | XP_016877700.1 | 24 | 3219 | |||
XM_017022212.1 | XP_016877701.1 | 23 | 3773 | |||
XM_017022213.1 | XP_016877702.1 | 23 | 3175 | |||
XM_017022214.1 | XP_016877703.1 | 24 | 3447 | |||
XM_017022215.1 | XP_016877704.1 | 24 | 3972 | |||
XM_017022216.1 | XP_016877705.1 | 23 | 4570 | |||
XM_017022217.1 | XP_016877706.1 | 22 | 4492 | |||
XM_017022218.1 | XP_016877707.1 | 24 | 4599 | |||
GenBank mRNA | AK097551.1 | - | 16 | 2290 | ||
BC144610.1 | - | 14 | 2035 | 9053141 | ||
BX647673.1 | - | 23 | 4517 |