Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145205.1 | NP_001138677.2 | 2 | 1750 | ||
GenBank mRNA | AK094972.1 | - | 1 | 285 | ||
AK309038.1 | - | 2 | 1091 | |||
AK309193.1 | - | 1 | 272 | |||
CR627300.1 | - | 2 | 946 | |||
CR749545.1 | - | 2 | 946 |