Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000149.3 | NP_000140.1 | 3 | 2343 | ||
NM_001097639.1 | NP_001091108.1 | 3 | 1995 | |||
NM_001097640.1 | NP_001091109.1 | 3 | 1941 | |||
NM_001097641.1 | NP_001091110.1 | 2 | 1802 | |||
XM_011527865.1 | XP_011526167.1 | 3 | 2321 | |||
XM_011527866.1 | XP_011526168.1 | 3 | 1982 | |||
XM_011527867.1 | XP_011526169.1 | 3 | 1951 | |||
GenBank mRNA | AF131913.1 | - | 2 | 1874 | ||
U27326.1 | - | 3 | 1934 | |||
U27327.1 | - | 3 | 1988 | |||
U27328.1 | - | 4 | 2337 | |||
X53578.1 | - | 2 | 1777 |