Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004339.2 | NP_001004339.2 | 5 | 1654 | ||
NM_001307931.1 | NP_001294860.1 | 4 | 902 | |||
XM_011541480.2 | XP_011539782.1 | 5 | 1429 | |||
XM_011541482.2 | XP_011539784.1 | 5 | 1429 | |||
XM_011541483.2 | XP_011539785.1 | 5 | 1429 | |||
XM_011541484.1 | XP_011539786.1 | 5 | 1429 | |||
XM_011541485.1 | XP_011539787.1 | 5 | 1429 | |||
XM_011541486.1 | XP_011539788.1 | 5 | 1429 | |||
XM_011541487.2 | XP_011539789.1 | 5 | 1429 | |||
XM_011541488.1 | XP_011539790.1 | 5 | 1429 | |||
GenBank mRNA | AY271826.1 | - | 4 | 893 |