Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004351.4 | NP_001004351.3 | 9 | 1651 | ||
XM_011516234.2 | XP_011514536.1 | 9 | 2554 | |||
XM_011516235.2 | XP_011514537.1 | 9 | 2554 | |||
GenBank mRNA | BC056606.1 | - | 9 | 1652 | 4838469 | |
BC100972.2 | - | 4 | 576 |