Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290216.2 | NP_001277145.1 | 4 | 567 | ||
GenBank mRNA | AK299756.1 | - | 1 | 142 | ||
DC376623.1 | - | 1 | 142 |