Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKI67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145966.1 | NP_001139438.1 | 12 | 7773 | ||
NM_002417.4 | NP_002408.3 | 13 | 8853 | |||
XM_006717864.3 | XP_006717927.2 | 2 | 6754 | |||
XM_011539818.2 | XP_011538120.1 | 10 | 8584 | |||
GenBank mRNA | X65550.1 | - | 13 | 8854 | ||
X65551.1 | - | 12 | 7774 |