Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002923.1 | NP_001002923.1 | 3 | 219 | ||
GenBank mRNA | AY672114.1 | - | 3 | 219 |