Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007189.1 | NP_001007190.1 | 1 | 2588 | ||
GenBank mRNA | AB073888.1 | - | 1 | 904 | ||
BC017422.1 | - | 1 | 180 | 4688423 | ||
BC041380.1 | - | 1 | 2592 |