Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYMSOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012716.2 | NP_001012734.2 | 1 | 290 | ||
GenBank mRNA | AK289986.1 | - | 1 | 219 | ||
BC028301.1 | - | 1 | 290 | 5141042 | ||
BM695182.1 | - | 1 | 230 |