Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024680.2 | NP_001019851.1 | 3 | 528 | ||
XM_005264407.2 | XP_005264464.1 | 3 | 530 | |||
XM_017004437.1 | XP_016859926.1 | 3 | 543 | |||
GenBank mRNA | BC089423.1 | - | 3 | 469 | 3103174 | |
BG435927.1 | - | 3 | 536 | |||
DQ904332.1 | - | 3 | 469 |