Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145194.1 | NP_001138666.1 | 2 | 276 | ||
XM_011525728.2 | XP_011524030.1 | 2 | 2603 |