Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129885.1 | NP_001123357.1 | 3 | 258 | ||
XM_011525115.2 | XP_011523417.1 | 3 | 2156 | |||
GenBank mRNA | BC157870.1 | - | 3 | 258 | 9057089 |