Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001444.2 | NP_001435.1 | 2 | 205 | ||
GenBank mRNA | AK226036.1 | - | 2 | 154 | ||
AK311856.1 | - | 2 | 152 | |||
BC019385.2 | - | 2 | 138 | 3862100 | ||
BC070303.1 | - | 1 | 152 | 6496304 | ||
BT007449.1 | - | 1 | 92 | |||
CN278427.1 | - | 2 | 325 | |||
M94856.1 | - | 2 | 140 |