Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256703.1 | NP_001243632.1 | 6 | 1137 | ||
NM_001256704.1 | NP_001243633.1 | 8 | 1290 | |||
NM_001267582.1 | NP_001254511.1 | 9 | 958 | |||
NM_002042.4 | NP_002033.2 | 7 | 1188 | |||
XM_017010689.1 | XP_016866178.1 | 8 | 778 | |||
GenBank mRNA | AK296124.1 | - | 6 | 729 | ||
AK298948.1 | - | 9 | 948 | |||
AK313609.1 | - | 8 | 1290 | |||
BC130344.1 | - | 7 | 742 | 40146375 | ||
M62400.1 | - | 7 | 761 |