Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040433.1 | NP_001035523.1 | 1, 2 | 107 | ||
GenBank mRNA | AY396740.1 | - | 1, 2 | 107 | ||
BC137530.1 | - | 1, 2 | 78 | 9021530 | ||
BC137531.1 | - | 1, 2 | 78 | 9021531 |