Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105562.2 | NP_001099032.1 | 24 | 3968 | ||
NM_006048.4 | NP_006039.2 | 23 | 3581 | |||
XM_005263422.1 | XP_005263479.1 | 25 | 3945 | |||
XM_011540488.2 | XP_011538790.1 | 24 | 3244 | |||
XM_011540489.2 | XP_011538791.1 | 25 | 3945 | |||
XM_011540490.1 | XP_011538792.1 | 25 | 3945 | |||
XM_017000035.1 | XP_016855524.1 | 22 | 3031 | |||
XM_017000036.1 | XP_016855525.1 | 24 | 3245 | |||
GenBank mRNA | AB014584.3 | - | 23 | 3472 | ||
AB028839.1 | - | 23 | 3589 | |||
AF043117.1 | - | 24 | 3365 | |||
AF331520.1 | - | 23 | 2940 | |||
AK292444.1 | - | 23 | 3063 | |||
AK304484.1 | - | 21 | 2568 | |||
BC093696.1 | - | 23 | 2971 | 7939541 | ||
BC143458.1 | - | 23 | 2971 | |||
BC143460.1 | - | 22 | 2856 | |||
JF289274.1 | - | 25 | 3433 |