Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080529.2 | NP_001073998.2 | 3, 4 | 406 | ||
XM_017012522.1 | XP_016868011.1 | 2, 3 | 414 |