Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRGX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276451.1 | NP_001263380.1 | 3 | 248 | ||
XM_011540089.2 | XP_011538391.1 | 2 | 166 | |||
GenBank mRNA | HY131276.1 | - | 3 | 248 |