Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039999.2 | NP_001035088.2 | 5 | 2204 | ||
XM_017024953.1 | XP_016880442.1 | 5 | 2288 | |||
GenBank mRNA | AK123558.1 | - | 3 | 2474 | ||
BC105988.1 | - | 1 | 188 | |||
BX402942.2 | - | 1 | 917 | |||
DY655105.1 | - | 1 | 274 |