Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143830.1 | NP_001137302.1 | 4 | 698 | ||
NM_005256.3 | NP_005247.1 | 3 | 489 | |||
NM_177553.2 | NP_808221.1 | 4 | 640 | |||
XM_011519971.2 | XP_011518273.1 | 4 | 698 | |||
XM_011519972.2 | XP_011518274.1 | 4 | 2693 | |||
XM_011519975.1 | XP_011518277.1 | 4 | 698 | |||
XM_011519978.2 | XP_011518280.1 | 4 | 698 | |||
XM_017017528.1 | XP_016873017.1 | 4 | 1390 | |||
XM_017017529.1 | XP_016873018.1 | 4 | 535 | |||
XM_017017530.1 | XP_016873019.1 | 4 | 728 | |||
XM_017017531.1 | XP_016873020.1 | 4 | 549 | |||
XM_017017532.1 | XP_016873021.1 | 4 | 698 | |||
GenBank mRNA | AK313734.1 | - | 4 | 704 | ||
BC013326.1 | - | 4 | 644 | |||
BC040470.1 | - | 4 | 640 | 4280621 | ||
CR450285.1 | - | 3 | 392 | |||
U95032.1 | - | 3 | 458 |