Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002139.3 | NP_002130.2 | 3, 4 | 429 | ||
NR_028477.1 | 3, 4 | 464 | ||||
GenBank mRNA | AK096015.1 | - | 2 | 335 | ||
AK304573.1 | - | 3, 4 | 409 | |||
AK309911.1 | - | 3, 4 | 384 | |||
AY464692.1 | - | 2, 3 | 219 | |||
BC006550.2 | - | 3, 4 | 365 | 3010123 | ||
BC007435.2 | - | 3, 4 | 365 | 3010123 | ||
BM147410.1 | - | 3, 4 | 429 | |||
DC337568.1 | - | 3, 4 | 409 | |||
Z23064.1 | - | 2, 3 | 230 |
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162536.2 | NP_001156008.1 | 3 | 935 | ||
NM_019610.5 | NP_062556.2 | 2 | 835 | |||
GenBank mRNA | AL832554.1 | - | 2 | 688 | ||
BC012942.1 | - | 2 | 494 | 3875565 |