Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000157.3 | NP_000148.2 | 5 | 678 | ||
NM_001005741.2 | NP_001005741.1 | 6 | 937 | |||
NM_001005742.2 | NP_001005742.1 | 6 | 918 | |||
NM_001171811.1 | NP_001165282.1 | 4 | 754 | |||
NM_001171812.1 | NP_001165283.1 | 4 | 528 | |||
GenBank mRNA | AK291911.1 | - | 6 | 916 | ||
AK298900.1 | - | 4 | 528 | |||
AK300186.1 | - | 4 | 598 | |||
AK300829.1 | - | 4 | 591 | |||
AK300876.1 | - | 6 | 918 | |||
AK301374.1 | - | 3 | 287 | |||
AK302000.1 | - | 4 | 503 | |||
AK311242.1 | - | 6 | 526 | |||
AK312502.1 | - | 6 | 734 | |||
AL547558.3 | - | 7 | 850 | |||
BC003356.1 | - | 5 | 617 | 2899915 | ||
BX648487.1 | - | 6 | 746 | |||
D13286.1 | - | 5 | 617 | |||
K02920.1 | - | 4 | 497 | |||
KJ690771.1 | - | 5 | 561 | |||
KJ690772.1 | - | 5 | 561 | |||
KJ690773.1 | - | 5 | 561 | |||
M16328.1 | - | 5 | 947 | |||
M19285.1 | - | 5 | 634 |
Gene Symbol: GBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002188.3 | 6 | 559 | |||
GenBank mRNA | AK295259.1 | - | 4 | 394 | ||
AK297653.1 | - | 4 | 369 | |||
BC000349.2 | - | 4 | 396 | |||
BC030240.1 | - | 6 | 438 | |||
BC092482.1 | - | 4 | 394 | |||
KJ690774.1 | - | 4 | 312 | |||
KJ690775.1 | - | 4 | 312 | |||
KJ690776.1 | - | 4 | 312 |