Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013627.2 | NP_001013649.2 | 8 | 3629 | ||
XM_011530934.2 | XP_011529236.1 | 8 | 8421 | |||
XM_017029473.1 | XP_016884962.1 | 8 | 7391 | |||
XM_017029474.1 | XP_016884963.1 | 8 | 3533 | |||
XM_017029475.1 | XP_016884964.1 | 4 | 6776 | |||
XM_017029476.1 | XP_016884965.1 | 4 | 2893 | |||
XM_017029477.1 | XP_016884966.1 | 4 | 7807 |