Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301089.1 | NP_001288018.1 | 8 | 989 | ||
NM_014758.2 | NP_055573.2 | 8 | 3003 | |||
XM_005271546.4 | XP_005271603.1 | 8 | 3002 | |||
XM_011542819.2 | XP_011541121.1 | 8 | 3002 | |||
XM_011542820.2 | XP_011541122.1 | 8 | 3002 | |||
XM_011542821.2 | XP_011541123.1 | 7 | 2883 | |||
XM_011542822.2 | XP_011541124.1 | 8 | 3002 | |||
XM_011542824.2 | XP_011541126.1 | 8 | 1907 | |||
XM_011542825.2 | XP_011541127.1 | 8 | 1169 | |||
XM_011542826.2 | XP_011541128.1 | 8 | 986 | |||
XM_011542827.2 | XP_011541129.1 | 7 | 867 | |||
XM_017017725.1 | XP_016873214.1 | 7 | 2858 | |||
XM_017017726.1 | XP_016873215.1 | 8 | 1166 | |||
GenBank mRNA | AF395843.1 | - | 8 | 2455 | ||
AK292457.1 | - | 8 | 3025 | |||
AK294731.1 | - | 5 | 517 | |||
AK304809.1 | - | 9 | 1009 | |||
AK308628.1 | - | 4 | 668 | |||
AK316414.1 | - | 8 | 953 | |||
AK316505.1 | - | 7 | 815 | |||
D87443.1 | - | 8 | 2983 | |||
DN990954.1 | - | 5 | 530 |