Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080752.3 | NP_542790.2 | 2 | 482 | ||
NR_037628.1 | 3 | 637 | ||||
GenBank mRNA | AK056641.1 | - | 2 | 447 | ||
AK302767.1 | - | 3 | 601 | |||
AK316214.1 | - | 3 | 600 | |||
BC027611.2 | - | 2 | 439 | 4829980 | ||
BX385024.2 | - | 2 | 440 | |||
DA520427.1 | - | 2 | 482 |