Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198542.1 | NP_001185471.1 | 19 | 2785 | ||
NM_001198543.1 | NP_001185472.1 | 19 | 2755 | |||
NM_001270974.2 | NP_001257903.1 | 19 | 2853 | |||
NM_017558.4 | NP_060028.2 | 19 | 2853 | |||
XM_006721206.2 | XP_006721269.1 | 19 | 2747 | |||
XM_011523146.2 | XP_011521448.1 | 20 | 2882 | |||
XM_011523147.1 | XP_011521449.1 | 20 | 2849 | |||
XM_011523148.1 | XP_011521450.1 | 20 | 2922 | |||
XM_011523151.1 | XP_011521453.1 | 19 | 2779 | |||
XM_017023346.1 | XP_016878835.1 | 19 | 3025 | |||
XM_017023347.1 | XP_016878836.1 | 7 | 1264 | |||
XM_017023348.1 | XP_016878837.1 | 7 | 1004 | |||
GenBank mRNA | AK022933.1 | - | 19 | 2787 | ||
AK057467.1 | - | 3 | 1157 | |||
AK299016.1 | - | 19 | 2742 | |||
AK299348.1 | - | 19 | 2785 | |||
AK308296.1 | - | 17 | 2372 | |||
AL122038.1 | - | 4 | 528 | |||
AL133042.1 | - | 4 | 528 |
Gene Symbol: HYDIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103556.1 | 14 | 2033 | |||
GenBank mRNA | AL833826.1 | - | 16 | 2229 | ||
AL834340.1 | - | 11 | 1523 |