Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024383.1 | NP_001019554.1 | 33 | 6141 | ||
NM_014903.5 | NP_055718.4 | 32 | 6075 | |||
XM_005269215.3 | XP_005269272.1 | 31 | 10055 | |||
XM_011538940.2 | XP_011537242.1 | 31 | 10035 | |||
XM_011538941.2 | XP_011537243.1 | 30 | 10013 | |||
XM_011538944.2 | XP_011537246.1 | 30 | 9543 | |||
XM_017020164.1 | XP_016875653.1 | 32 | 10123 | |||
XM_017020165.1 | XP_016875654.1 | 31 | 10081 | |||
XM_017020166.1 | XP_016875655.1 | 33 | 6130 | |||
XM_017020167.1 | XP_016875656.1 | 32 | 6061 | |||
XM_017020168.1 | XP_016875657.1 | 32 | 9654 | |||
XM_017020169.1 | XP_016875658.1 | 31 | 9611 | |||
XM_017020170.1 | XP_016875659.1 | 29 | 9521 | |||
XM_017020171.1 | XP_016875660.1 | 25 | 9437 | |||
XM_017020172.1 | XP_016875661.1 | 22 | 4946 | |||
XM_017020173.1 | XP_016875662.1 | 24 | 8946 | |||
XM_017020174.1 | XP_016875663.1 | 23 | 8904 | |||
XM_017020175.1 | XP_016875664.1 | 23 | 4759 | |||
GenBank mRNA | AB023155.2 | - | 17 | 2585 | ||
AF397731.1 | - | 31 | 5659 | |||
AJ488201.1 | - | 33 | 6013 | |||
AK298525.1 | - | 15 | 1768 |