Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024383.1 | NP_001019554.1 | 38 | 6925 | ||
NM_014903.5 | NP_055718.4 | 37 | 6859 | |||
XM_005269215.3 | XP_005269272.1 | 36 | 10839 | |||
XM_011538940.2 | XP_011537242.1 | 36 | 10819 | |||
XM_011538941.2 | XP_011537243.1 | 35 | 10797 | |||
XM_011538944.2 | XP_011537246.1 | 35 | 10327 | |||
XM_017020164.1 | XP_016875653.1 | 37 | 10907 | |||
XM_017020165.1 | XP_016875654.1 | 36 | 10865 | |||
XM_017020166.1 | XP_016875655.1 | 38 | 6914 | |||
XM_017020167.1 | XP_016875656.1 | 37 | 6845 | |||
XM_017020168.1 | XP_016875657.1 | 37 | 10438 | |||
XM_017020169.1 | XP_016875658.1 | 36 | 10395 | |||
XM_017020170.1 | XP_016875659.1 | 34 | 10305 | |||
XM_017020171.1 | XP_016875660.1 | 30 | 10221 | |||
XM_017020172.1 | XP_016875661.1 | 27 | 5730 | |||
XM_017020173.1 | XP_016875662.1 | 29 | 9730 | |||
XM_017020174.1 | XP_016875663.1 | 28 | 9688 | |||
XM_017020175.1 | XP_016875664.1 | 28 | 5543 | |||
GenBank mRNA | AB023155.2 | - | 22 | 3369 | ||
AF397731.1 | - | 36 | 6443 | |||
AJ488201.1 | - | 38 | 6797 | |||
AK294988.1 | - | 3 | 354 |