Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278275.1 | NP_001265204.1 | 4 | 712 | ||
NM_001278276.1 | NP_001265205.1 | 4 | 774 | |||
NM_001278277.1 | NP_001265206.1 | 5 | 845 | |||
NM_001278278.1 | NP_001265207.1 | 2 | 586 | |||
NM_198256.3 | NP_937987.2 | 3 | 641 | |||
NM_212540.2 | NP_997705.1 | 3 | 565 | |||
NR_103490.1 | 3 | 641 | ||||
XM_017003547.1 | XP_016859036.1 | 3 | 625 | |||
XM_017003548.1 | XP_016859037.1 | 4 | 712 | |||
XM_017003549.1 | XP_016859038.1 | 2 | 570 | |||
GenBank mRNA | AF041381.1 | - | 3 | 327 | ||
AF059292.1 | - | 3 | 343 | |||
AK096197.1 | - | 5 | 547 | |||
AK223159.1 | - | 3 | 314 | |||
AK223407.1 | - | 3 | 620 | |||
AK290413.1 | - | 3 | 613 | |||
AK293504.1 | - | 3 | 613 | |||
AU132447.1 | - | 3 | 615 | |||
AU142999.1 | - | 2 | 580 | |||
AY083996.1 | - | 4 | 692 | |||
AY551346.1 | - | 3 | 343 | |||
AY551347.1 | - | 4 | 414 | |||
AY551348.1 | - | 4 | 476 | |||
AY551349.1 | - | 5 | 547 | |||
AY551350.1 | - | 2 | 288 | |||
AY551351.1 | - | 3 | 267 | |||
BC008348.1 | - | 3 | 600 | 3535798 | ||
BC062610.1 | - | 3 | 534 | |||
BC107740.1 | - | 3 | 558 | 6494695 | ||
BI772069.1 | - | 2 | 466 | |||
BU076470.1 | - | 4 | 452 | |||
KU178035.1 | - | 3 | 343 | |||
KU178036.1 | - | 2 | 288 |