Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010110.1 | XP_016865599.1 | 12 | 1614 | ||
XM_017010111.1 | XP_016865600.1 | 11 | 1441 | |||
XM_017010112.1 | XP_016865601.1 | 11 | 2018 | |||
XM_017010113.1 | XP_016865602.1 | 12 | 2140 | |||
XM_017010114.1 | XP_016865603.1 | 12 | 4901 | |||
XM_017010115.1 | XP_016865604.1 | 11 | 1966 | |||
XM_017010116.1 | XP_016865605.1 | 10 | 2366 | |||
XM_017010117.1 | XP_016865606.1 | 10 | 2201 | |||
XM_017010120.1 | XP_016865609.1 | 11 | 3314 | |||
XM_017010122.1 | XP_016865611.1 | 11 | 3311 | |||
XM_017010123.1 | XP_016865612.1 | 10 | 2115 | |||
XM_017010124.1 | XP_016865613.1 | 11 | 2125 | |||
XM_017010125.1 | XP_016865614.1 | 7 | 1690 |