Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN20 SIRNA Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases:
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
Chromosome Location:
Chr.10: 46911134 - 47003834 on Build GRCh38
UniGene ID:
Hs.440733
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s57579
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPN20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042357.4
NP_001035816.1
11
2311
NM_001042358.4
NP_001035817.1
10
2169
NM_001042359.3
NP_001035818.1
8
1743
NM_001042360.3
NP_001035819.1
5
1620
NM_001042361.4
NP_001035820.1
8
1618
NM_001042362.3
NP_001035821.1
6
1375
NM_001042363.4
NP_001035822.1
10
2154
NM_001042364.4
NP_001035823.1
7
1508
NM_001042365.3
NP_001035824.1
4
1167
NM_001320681.1
NP_001307610.1
9
2059
NM_001320682.1
NP_001307611.1
5
1265
NM_001320683.1
NP_001307612.1
9
1775
NM_001320684.1
NP_001307613.1
8
1684
NM_001320685.1
NP_001307614.1
10
2231
NM_001320686.1
NP_001307615.1
9
2074
NM_001320688.1
NP_001307617.1
7
1648
NM_001320689.1
NP_001307618.1
6
1416
NM_001320690.1
NP_001307619.1
5
1280
NM_001320691.1
NP_001307620.1
8
1983
NM_015605.8
NP_056420.3
9
1760
XM_011539605.2
XP_011537907.1
11
2455
XM_011539606.2
XP_011537908.1
11
3547
XM_011539607.2
XP_011537909.1
11
2446
XM_011539610.2
XP_011537912.1
8
1929
XM_011539611.2
XP_011537913.1
7
1929
XM_017016042.1
XP_016871531.1
11
2392
XM_017016044.1
XP_016871533.1
10
2212
XM_017016045.1
XP_016871534.1
5
3531
XM_017016046.1
XP_016871535.1
10
2074
XM_017016049.1
XP_016871538.1
8
1805
XM_017016050.1
XP_016871539.1
6
1672
XM_017016053.1
XP_016871542.1
8
1648
XM_017016054.1
XP_016871543.1
8
1661
XM_017016055.1
XP_016871544.1
8
1750
XM_017016056.1
XP_016871545.1
7
1656
XM_017016057.1
XP_016871546.1
8
1966
XM_017016058.1
XP_016871547.1
6
1692
XM_017016059.1
XP_016871548.1
7
1650
XM_017016060.1
XP_016871549.1
7
1496
XM_017016061.1
XP_016871550.1
6
1446
XM_017016062.1
XP_016871551.1
6
1566
XM_017016063.1
XP_016871552.1
7
1537
XM_017016064.1
XP_016871553.1
5
1289
XM_017016065.1
XP_016871554.1
6
1339
GenBank mRNA
AL050040.1
-
9
2017
BU567804.1
-
1
228
BX648913.1
-
4
1168
CR933668.1
-
5
1233
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0