Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL26L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317980.1 | NP_001304909.1 | 3, 4 | 428 | ||
NM_001317981.1 | NP_001304910.1 | 3, 4 | 402 | |||
NM_001317982.1 | NP_001304911.1 | 3, 4 | 384 | |||
NM_016093.3 | NP_057177.1 | 3, 4 | 374 | |||
XM_005265921.3 | XP_005265978.1 | 3, 4 | 425 | |||
XM_011534565.2 | XP_011532867.1 | 3, 4 | 989 | |||
XM_017009519.1 | XP_016865008.1 | 3, 4 | 416 | |||
XM_017009520.1 | XP_016865009.1 | 3, 4 | 347 | |||
GenBank mRNA | AF083248.1 | - | 3, 4 | 348 | ||
AK128872.1 | - | 1 | 342 | |||
BC017360.2 | - | 3, 4 | 362 | 4551093 | ||
BC070192.1 | - | 3, 4 | 347 | 6727802 | ||
BI838595.1 | - | 402 | ||||
BQ278240.1 | - | 3, 4 | 375 | |||
BU193755.1 | - | 3, 4 | 408 | |||
BU569345.1 | - | 3, 4 | 387 |