Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143989.2 | NP_001137461.1 | 5 | 760 | ||
XM_011540763.1 | XP_011539065.1 | 5 | 746 | |||
GenBank mRNA | AK302318.1 | - | 5 | 755 | ||
AK316438.1 | - | 5 | 746 | |||
BC125162.1 | - | 5 | 721 |
Gene Symbol: NBPF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143987.1 | NP_001137459.1 | 5 | 760 | ||
NM_001143988.1 | NP_001137460.1 | 5 | 760 | |||
XM_005271144.3 | XP_005271201.1 | 5 | 884 | |||
XM_011542012.2 | XP_011540314.1 | 10 | 2213 | |||
XM_011542014.2 | XP_011540316.1 | 5 | 1522 | |||
XM_017002148.1 | XP_016857637.1 | 10 | 1348 | |||
GenBank mRNA | BC125161.1 | - | 5 | 721 |