Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000854.4 | NP_000845.2 | 5 | 1080 | ||
NM_001302670.1 | NP_001289599.1 | 5 | 1038 | |||
NR_126442.1 | 6 | 1072 | ||||
NR_126445.1 | 5 | 1080 | ||||
NR_126446.1 | 6 | 1142 | ||||
GenBank mRNA | CD013993.1 | - | 6 | 950 | ||
L38503.1 | - | 5 | 910 |
Gene Symbol: GSTT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080843.3 | NP_001074312.1 | 5 | 960 | ||
XM_005261715.4 | XP_005261772.1 | 5 | 938 | |||
GenBank mRNA | BC002415.1 | - | 5 | 965 | 3164017 | |
BC071700.1 | - | 5 | 932 | 4932043 |