Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCP4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102566.1 | NP_001096036.1 | 3 | 361 | ||
XM_017002154.1 | XP_016857643.1 | 3 | 335 | |||
GenBank mRNA | BC028905.2 | - | 3 | 361 | ||
BC067884.1 | - | 3 | 339 | |||
BC147003.1 | - | 3 | 262 | 9056931 | ||
BC147004.1 | - | 3 | 262 | 9056932 | ||
CN283207.1 | - | 3 | 327 |