Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROCK1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033770.1 | 3 | 525 | |||
GenBank mRNA | BC041849.1 | - | 3 | 525 | ||
DB095042.1 | - | 3 | 162 |