Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002458.2 | NP_002449.2 | 6 | 709 | ||
GenBank mRNA | AF086604.1 | - | 6 | 650 | ||
AK310939.1 | - | 6 | 708 |