Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016458.3 | NP_057542.2 | 6 | 1820 | ||
GenBank mRNA | AF151022.1 | - | 3 | 1210 | ||
AK098226.1 | - | 7 | 1514 | |||
BC003035.1 | - | 6 | 1820 | 2822563 | ||
BC009915.2 | - | 6 | 1820 | 2822563 | ||
BC015471.1 | - | 7 | 1351 |